Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAAACCAGGAGCTGTATTTGTGA[C/T]TCATTTTCTAACAACACCAACATTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mad1l1 PubMed Links |
Mad1l1 - MAD1 mitotic arrest deficient 1-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010752.3 | Intron | NP_034882.2 |