Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCGCTATAGACAGCATAGTACTG[A/G]GGTTAAGACAAGCTTTGGTTCCCAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nr6a1 PubMed Links |
Nr6a1 - nuclear receptor subfamily 6, group A, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159548.1 | Intron | NP_001153020.1 | ||||
NM_001159549.1 | Intron | NP_001153021.1 | ||||
NM_010264.4 | Intron | NP_034394.1 |