Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGCCCTGGCCCCGTGTGACCTG[C/T]GACTTGGGATAGATGACCGTAGGGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slco4a1 PubMed Links |
Slco4a1 - solute carrier organic anion transporter family, member 4a1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_148933.1 | Intron | NP_683735.1 |