Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mdh1 PubMed Links |
Mdh1 - malate dehydrogenase 1, NAD (soluble) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008618.3 | Intron | NP_032644.3 |
Wdpcp - WD repeat containing planar cell polarity effector | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |