Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCATCTTACAGTACATTCACTACA[A/G]TGAGGTATATGAGGCCATCAGCATC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Wdpcp PubMed Links |
Wdpcp - WD repeat containing planar cell polarity effector | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145425.3 | 1517 | Missense Mutation | NP_663400.2 |