Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCGTAGCGGTCTTCCGGATCTG[C/T]GGACTGTCCATCCGGTTCTTCAAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Srgap2 PubMed Links |
Srgap2 - SLIT-ROBO Rho GTPase activating protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081011.2 | 3469 | Silent Mutation | NP_001074480.2 |