Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTACTTTGCGGCTGTTATCATAC[A/G]AAGTGGATACAAAAGTAGAACTTCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lrrk2 PubMed Links |
Lrrk2 - leucine-rich repeat kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025730.3 | Intron | NP_080006.3 |