Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTGAGTGATCAGAGCTGCTTAT[A/G]CCATCCAAAATCCTTATCTAAAGTA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cand1 PubMed Links |
Cand1 - cullin associated and neddylation disassociated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_027994.1 | 7842 | UTR 3 | NP_082270.1 |