Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCTCTTGCTTCCTCTCAGGCAC[A/G]ACAATGTAGGAAACTTAGCCAGGCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Lmx1b PubMed Links |
Lmx1b - LIM homeobox transcription factor 1 beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010725.2 | Intron | NP_034855.2 |