Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTCTACATGCTCAATGGACTCAC[A/G]ACTAATATGTCCCAGCTTCACAGCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Bend3 PubMed Links |
Bend3 - BEN domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199028.2 | 856 | Silent Mutation | NP_950193.1 |