Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTAAATGGAAAAATCCCAGCTAG[A/G]CATGGTCACCAGAGCGCTATGTCCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tpd52 PubMed Links |
Tpd52 - tumor protein D52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025261.1 | Intron | NP_001020432.1 | ||||
NM_001025262.1 | Intron | NP_001020433.1 | ||||
NM_001025263.1 | Intron | NP_001020434.1 | ||||
NM_001025264.1 | Intron | NP_001020435.1 | ||||
NM_009412.2 | Intron | NP_033438.1 |