Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGGATCATAACACACCCAGAAAA[A/G]CCTGCAGGAGGGGTCTGTCTTGTTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rit2 PubMed Links |
Rit2 - Ras-like without CAAX 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009065.2 | Intron | NP_033091.1 |