Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGATCACACATATTTGGATATAA[A/G]GTATATACGCTTAGGAATAATTGCC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rspo2 PubMed Links |
Rspo2 - R-spondin 2 homolog (Xenopus laevis) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172815.3 | Intron | NP_766403.1 |