Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAACGGTTTTATTTTTCCTCCA[C/T]TGGAAGTTTGAAGATGTACTCTTTG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kcnq5 PubMed Links |
Kcnq5 - potassium voltage-gated channel, subfamily Q, member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160139.1 | Intron | NP_001153611.1 | ||||
NM_023872.3 | Intron | NP_076361.1 |