Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGACTGCTTCATCTGCTGTATC[C/T]TATCCCACGGTGACAAGGGTGTCGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Casp8 PubMed Links |
Casp8 - caspase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080126.1 | 1285 | Silent Mutation | NP_001073595.1 | |||
NM_001277926.1 | 1285 | Silent Mutation | NP_001264855.1 | |||
NM_009812.2 | 1285 | Silent Mutation | NP_033942.1 |