Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Farp2 PubMed Links |
Farp2 - FERM, RhoGEF and pleckstrin domain protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Stk25 - serine/threonine kinase 25 (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021537.3 | Intron | NP_067512.3 |