Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAAGGTCTAGAATCAGCAAACAG[C/T]GAAGCTATGCTGCATACTGGGGAGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fmn2 PubMed Links |
Fmn2 - formin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019445.2 | Intron | NP_062318.2 |