Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAACTCAGACATTGGACTGAGAGA[T/C]ATGGGAAGGTCACTACCTACTGTAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Anxa5 PubMed Links |
Anxa5 - annexin A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009673.2 | 1486 | UTR 3 | NP_033803.1 |