Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dnajb14 PubMed Links |
Dnajb14 - DnaJ (Hsp40) homolog, subfamily B, member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033155.1 | Intron | NP_001028327.1 |
H2afz - H2A histone family, member Z | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |