Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAGAATAGAGAACTTAATTTAC[A/G]GTACTTGGTTATTGTGATGCGGAAT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Fezf1 PubMed Links |
Fezf1 - Fez family zinc finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_028462.1 | Intron | NP_082738.1 |