Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hoxa10 PubMed Links |
Hoxa10 - homeobox A10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Hoxa11 - homeobox A11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_010450.3 | 1251 | UTR 3 | NP_034580.1 |
Hoxa11os - homeobox A11, opposite strand | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |