Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGCGCCGGCGAATGAAGAAACC[G/A]GCACCACAGCAGAACAGCACACCCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Vopp1 PubMed Links |
Vopp1 - vesicular, overexpressed in cancer, prosurvival protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_146168.1 | 376 | Silent Mutation | NP_666280.1 |