Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTACCATCACAGCTAATAAAACC[C/T]CAGTTAGATTCCTAGAGCCCACATG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Utrn PubMed Links |
Utrn - utrophin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_011682.4 | Intron | NP_035812.3 |