Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGCAAATGCCTTTGAGCATCGG[A/G]GAGAATACCAAAACACTTGAACTCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Grip1 PubMed Links |
Grip1 - glutamate receptor interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277292.1 | Intron | NP_001264221.1 | ||||
NM_001277293.1 | Intron | NP_001264222.1 | ||||
NM_001277294.1 | Intron | NP_001264223.1 | ||||
NM_001277295.1 | Intron | NP_001264224.1 | ||||
NM_028736.2 | Intron | NP_083012.1 | ||||
NM_130891.2 | Intron | NP_570961.1 | ||||
NM_133442.2 | Intron | NP_597699.1 |