Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGATCTGGAGTCTATGCCAGC[G/A]TCTCTCAGGACTTTCAGGCTTATGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Tpo PubMed Links |
Tpo - thyroid peroxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009417.2 | 3086 | UTR 3 | NP_033443.1 |