Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTTTCTGATAAGAGGACTCTAC[A/T]GTCTACCGGCTACCCCATCCCAATC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Bmp6 PubMed Links |
Bmp6 - bone morphogenetic protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007556.2 | Intron | NP_031582.1 |