Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATAGTTTGGGTCCTTTTGTGCTC[G/A]TCTAGACTATCCTCTGCTTCCGTGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Nadk2 PubMed Links |
Nadk2 - NAD kinase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040395.4 | Intron | NP_001035485.2 | ||||
NM_001085410.2 | Intron | NP_001078879.1 | ||||
NM_001286253.1 | Intron | NP_001273182.1 | ||||
NM_001286255.1 | Intron | NP_001273184.1 |