Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATAAGTGAAATTAGAACATCAGT[C/T]ACTATACTGAATAATCTCCAATTAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hrsp12 PubMed Links |
Hrsp12 - heat-responsive protein 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008287.3 | 708 | UTR 3 | NP_032313.2 |