Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTTGGGAATTTTTTCTAAGAGT[G/A]CCAGTGTCAACTGTGACGTTTTTAA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Vps13b PubMed Links |
Vps13b - vacuolar protein sorting 13B (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177151.3 | Intron | NP_796125.2 |