Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Poldip3 PubMed Links |
Poldip3 - polymerase (DNA-directed), delta interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178627.3 | 2825 | UTR 3 | NP_848742.1 |
Rrp7a - ribosomal RNA processing 7 homolog A (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Serhl - serine hydrolase-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |