Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mrps18a PubMed Links |
Mrps18a - mitochondrial ribosomal protein S18A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_026768.3 | Intron | NP_081044.1 |
Rsph9 - radial spoke head 9 homolog (Chlamydomonas) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_029338.3 | Intron | NP_083614.1 |