Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGATGGGAAATAAGAACATGGCA[T/A]TTACTCCAAAAGTCATTTGCTTGGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Arhgap28 PubMed Links |
Arhgap28 - Rho GTPase activating protein 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_172964.4 | Intron | NP_766552.3 |