Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGCAAACCACTAGGAAGGGCTG[A/T]TGGCACATATACCTGTGTTTTAAGG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rock1 PubMed Links |
Rock1 - Rho-associated coiled-coil containing protein kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_009071.2 | Intron | NP_033097.1 |