Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACTTCCACATGAGCGGTGTTGA[T/G]GTCCGTCATAAGGAAACACACAGGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ostf1 PubMed Links |
Ostf1 - osteoclast stimulating factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017375.3 | Intron | NP_059071.1 |