Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGCTACCTTCCAGTTGATGGAG[A/G]CTTGAGAGACTCAGCCTGGCCTGTT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Kank1 PubMed Links |
Kank1 - KN motif and ankyrin repeat domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181404.5 | Intron | NP_852069.4 |