Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCACAGGGGAAGGTACTGACTGC[A/G]CCCACCCAAATGTCAAACAAATGTC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Cdkn2b PubMed Links |
Cdkn2b - cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007670.4 | 990 | UTR 3 | NP_031696.1 |