Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTGCAAGCCTGGGTGGCTTGCT[C/T]ATGGTGTCTGTGGATTTTTTGATGT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Rdh10 PubMed Links |
Rdh10 - retinol dehydrogenase 10 (all-trans) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133832.3 | 1588 | UTR 3 | NP_598593.1 |