Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dnm3 PubMed Links |
Dnm3 - dynamin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001038619.1 | Intron | NP_001033708.1 | ||||
NM_172646.2 | Intron | NP_766234.1 |
Dnm3os - dynamin 3, opposite strand | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Mir199a-2 - microRNA 199a-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Mir214 - microRNA 214 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |