Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Mir3962 PubMed Links |
Mir3962 - microRNA 3962 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Plxna2 - plexin A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008882.2 | 6742 | UTR 3 | NP_032908.2 |