Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCATGTCCTGGGCTGTTTCTCCC[C/T]GGAGGAGCAGGAGCTACTGTCCCCA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
1700019L03Rik PubMed Links |
1700019L03Rik - RIKEN cDNA 1700019L03 gene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025619.1 | 524 | Intron | NP_079895.1 |
Ptrh1 - peptidyl-tRNA hydrolase 1 homolog (S. cerevisiae) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178595.3 | 524 | Missense Mutation | NP_848710.1 |
Ttc16 - tetratricopeptide repeat domain 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |