Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTCCCTTTATCTTTTTTATAA[A/G]ACTCAATCGTCAGTTCTATACTAGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Slc24a3 PubMed Links |
Slc24a3 - solute carrier family 24 (sodium/potassium/calcium exchanger), member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_053195.2 | Intron | NP_444425.1 |