Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATATGCAGACCTGCCGGGAGAA[C/T]TACACATTATCGAACTGGAAAAGGA
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Inadl PubMed Links |
Inadl - InaD-like (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005784.1 | 410 | Intron | NP_001005784.1 | |||
NM_001005787.1 | 410 | Silent Mutation | NP_001005787.1 | |||
NM_007704.2 | 410 | Silent Mutation | NP_031730.1 | |||
NM_172696.2 | 410 | Silent Mutation | NP_766284.2 |