Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Ccdc162 PubMed Links |
Ccdc162 - coiled-coil domain containing 162 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Cd164 - CD164 antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016898.2 | 2550 | UTR 3 | NP_058594.1 |