Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAGAAAGACTGCAGTAAGTAGC[C/T]GGTCCCCTCTCACCTGCACAGATAG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Egfr PubMed Links |
Egfr - epidermal growth factor receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007912.4 | Intron | NP_031938.1 | ||||
NM_207655.2 | Intron | NP_997538.1 |