Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCTATCAATCAGTGTTTTTGTGG[C/T]TAGTGAAATCATGTTAATAACTTAC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Hdac9 PubMed Links |
Hdac9 - histone deacetylase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271386.1 | Intron | NP_001258315.1 | ||||
NM_024124.3 | Intron | NP_077038.2 |