Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTCCCAGCATCCCTCTCCAACCT[C/T]CCCCTTCAGAAGCCTACGAGTGTCT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Npas3 PubMed Links |
Npas3 - neuronal PAS domain protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013780.2 | Intron | NP_038808.2 |