Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAATAATATAAATTGTTAAGAATT[A/G]AAGAATTCTGTTTCCTGGAATGAGC
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Bicd2 PubMed Links |
Bicd2 - bicaudal D homolog 2 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039179.2 | 5903 | UTR 3 | NP_001034268.1 | |||
NM_001039180.2 | 5903 | UTR 3 | NP_001034269.1 | |||
NM_029791.4 | 5903 | UTR 3 | NP_084067.1 |