Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Dock5 PubMed Links |
Dock5 - dedicator of cytokinesis 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Gnrh1 - gonadotropin releasing hormone 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_008145.2 | Intron | NP_032171.1 |
Kctd9 - potassium channel tetramerisation domain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |