Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTTCCCATGTAAATAGGGCCTT[G/T]AAATGTAAATAACTTTAATAAAACG
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Myc PubMed Links |
Myc - myelocytomatosis oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177352.1 | 2248 | UTR 3 | NP_001170823.1 | |||
NM_001177353.1 | 2248 | UTR 3 | NP_001170824.1 | |||
NM_001177354.1 | 2248 | UTR 3 | NP_001170825.1 | |||
NM_010849.4 | 2248 | UTR 3 | NP_034979.3 |