Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGGTGAAAAGAACCGTGAGTAG[C/G]CAGATGAATTTACTGTTGGTTCATT
Species: |
Mouse |
dbSNP Submissions: |
NA
|
Phenotype: |
|
Literature Links: |
Robo1 PubMed Links |
Robo1 - roundabout homolog 1 (Drosophila) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019413.2 | Intron | NP_062286.2 |